

(±)-3,5,5-Trimethyl-1-hexanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:3452-97-9
:
Formula:(CH₃)₃CCH₂CH(CH₃)CH₂CH₂OH
MDL Number:MFCD00036138
Boiling Pt:193…194 °C (1013 hPa)
Melting Pt:–70 °C
Density:0.83 g/cm³ (20 °C)
Flash Pt:80 °C (closed cup)
EINECS:222-376-7
UN:3082
ADR:9,III
:M-Clarity™ quality level = MQ100
:144.26 g/mol
:GHS08
:GHS09
Storage Temperature:Ambient
Frequently Bought Together