
(±)-2-Ethyl-1-hexanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:104-76-7
:
Formula:CH₃(CH₂)₃CH(C₂H₅)CH₂OH
MDL Number:MFCD00004746
:130.23 g/mol
Merck Index:13,03842
Boiling Pt:183 °C (1013 hPa)
Melting Pt:–76 °C
Density:0.83 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Flash Pt:81 °C
Storage Temperature:Ambient
EINECS:203-234-3
REACH:01-2119487289-20
:GHS07
Frequently Bought Together