(±)-1,2-Propanediol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:57-55-6
:
Formula:CH₃CH(OH)CH₂OH
MDL Number:MFCD00064272
:76.1 g/mol
Merck Index:13,07947
Boiling Pt:187.6 °C (1013 hPa)
Melting Pt:–59 °C
Density:1.04 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:99 °C
Storage Temperature:Ambient
EINECS:200-338-0
Frequently Bought Together