Order Entry
Czech Republic
ContactUsLinkComponent
(±)-1,2-Butanediol, Sigma-Aldrich®
(±)-1,2-Butanediol, Sigma-Aldrich®
Catalog # SIAL177652-250G
Supplier:  Merck
CAS Number:  
(±)-1,2-Butanediol, Sigma-Aldrich®
Catalog # SIAL177652-250G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :90.12 g/mol
Boiling Pt:193 °C (1013 hPa)
Melting Pt:–50 °C
Density:1.0189 g/cm³ (0 °C)
 :M-Clarity™ quality level = MQ100
EINECS:209-527-2
CAS Number:584-03-2
 :26171-83-5
Formula:C₄H₁₀O₂
MDL Number:MFCD00004570