


1,4-Dioxane, extra dry over Molecular Sieve 99,5% stabilised, AcroSeal®
:123-91-1
:
:C₄H₈O₂
:MFCD00006571
:88.11 g/mol
:13,03330
:101 °C (760 mmHg)
:12 °C
:1.034 g/cm³
:12 °C
:Ambient
:204-661-8
:1165
:3,II
:GHS02
:GHS07
:GHS08
- Appearance:Clear colourless liquid
- Infrared spectrum:Conforms
- Water:≤0,005 %
- GC:≥99,5 %
- Stabilizer:2 to 5 ppm BHT
- Acidity (CH3COOH):≤0,002 %
- Peroxides:≤0,005 % (as H₂O₂)
Frequently Bought Together