
1,3-Cyclohexanedione 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:504-02-9
:
Formula:C₆H₈O₂
MDL Number:MFCD00001585
:112.13 g/mol
Merck Index:13,03200
Boiling Pt:235 °C (1013 hPa)
Melting Pt:>101 °C
Density:1,25 g/cm³ (20 °C)
EINECS:207-980-0
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Light yellow to beige Powder
- Infrared spectrum:Conforms
- Melting point:≥101 °C
- Titration Argentometric:≤1 % Sodium chloride
- Water:≤0.5 %
- Titration with NaOH:≥96.0 %
Specification Test Results
Frequently Bought Together