



3-Buten-2-one stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:78-94-4
:
Formula:C₄H₆O
MDL Number:MFCD00008777
:70.09 g/mol
Merck Index:12,06214
Boiling Pt:81.4 °C (1013 hPa)
Melting Pt:–7 °C
Density:0.8636 g/cm³ (20 °C)
Storage Temperature:Freezer
ADR:6.1 (3, 8),I
:M-Clarity™ quality level = MQ200
Flash Pt:–7 °C (closed cup)
:Stabilised with hydroquinone (HQ)
EINECS:201-160-6
UN:1251
:GHS02
:GHS06
:GHS05
:GHS09
Related Information
Certificates
Search by Lot Numbermsds_805901
SDS
ProductPage_805901 ProductPage_805901
Product specification
Frequently Bought Together