



3-Buten-2-one tech. 90% stabilised, Technical Grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78-94-4
:
Formula:C₄H₆O
MDL Number:MFCD00008777
:70.09 g/mol
Merck Index:14,6134
Boiling Pt:32…34 °C (60 mmHg)
Melting Pt:–7 °C
Density:0.856
Flash Pt:–6 °C (21 °F)
UN:1251
ADR:6.1,I
:Stabilised with hydroquinone (HQ) 0.5 %
EINECS:201-160-6
:GHS02
:GHS06
:GHS05
:GHS09
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together