Supelco® Organic Reference Standard, 1,2-Dimethoxyethane
Catalog # 72405-5ML-F
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mL
- Standards:Analytical standard
- CAS No.:110-71-4
- Description:1,2-Dimethoxyethane
- Seq:02
- Storage conditions:2...30 °C
- Hazard class description:H225 - H315 - H332 - H360FD
- Shipping conditions:STD
- MDL No.:MFCD00008502
- M-Clarity quality level:MQ100
- Synonym(s):mono-Glyme; Dimethylglycol; Ethylene glycol dimethyl ether; Monoglyme
- Melting point:–58 °C (lit.)
- Formula:CH₃OCH₂CH₂OCH₃
- Molecular weight:90,12
- Density:0,867 g/ml at 25 °C (lit.)
- Flash point:5 °C - Closed cup
- Boiling point:85 °C (lit.)
Specifications
About this item
- GHS signal: GHS02, GHS08, GHS07
Caution: P202 - P210 - P233 - P303 + P361 + P353 - P304 + P340 + P312 - P308 + P313