Order Entry
Czech Republic
ContactUsLinkComponent
L(+)-Cysteine hydrochloride, anhydrous 98%
Irritant
L(+)-Cysteine hydrochloride, anhydrous 98%
Catalog # L06328.14
CAS Number:  
Irritant
L(+)-Cysteine hydrochloride, anhydrous 98%
Catalog # L06328.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

About this item

CAS Number:52-89-1
 :
Formula:HSCH₂CH(NH₂)COOH·HCl
MDL Number:MFCD00064552
 :157.62 g/mol
Merck Index:14,2781
Melting Pt:∼ 180 °C (dec.)
EINECS:200-157-7
 :GHS07
Storage Temperature:Ambient
MDL Number:MFCD00064553