Order Entry
Czech Republic
ContactUsLinkComponent
Tween® 85 (Polysorbate) for synthesis, Sigma-Aldrich®
Tween® 85 (Polysorbate) for synthesis, Sigma-Aldrich®
Catalog # 8.22188.0500
Supplier:  Merck
CAS Number:  
Tween® 85 (Polysorbate) for synthesis, Sigma-Aldrich®
Catalog # 8.22188.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Vapor Pressure:
    1.4 hPa (20 °C)
  • Flash Point:
    149 °C DIN 51758
  • Density:
    1.03 g/cm3 (20 °C)

Specification Test Results

About this item

 :
Density:1.02 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
CAS Number:9005-70-3
MDL Number:MFCD01779641