Order Entry
Czech Republic
ContactUsLinkComponent
 
Tween® 60 (Polysorbate) for synthesis, Sigma-Aldrich®
 :  Merck
 
Tween® 60 (Polysorbate) for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Hydroxyl value:
    81 - 96
  • Saponification value:
    45 - 55
  • Identity (IR):
    passes test
  • Density:
    1.1 g/cm3 (20 °C)
  • pH:
    5.5 - 7.7 (50 g/l, H2O, 25 °C)
  • Viscosity kinematic:
    170 - 320 mm2/s (40 °C)

 

Frequently Bought Together