Tween® 20 (Polysorbate), Sigma-Aldrich®
Catalog # SIAL93773-1KG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:1227.53 g/mol
Density:1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:500-018-3
CAS Number:9005-64-5
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986