Order Entry
Czech Republic
ContactUsLinkComponent
Polysorbate 20, clear cell culture reagent
Polysorbate 20, clear cell culture reagent
Supplier:  MP Biomedicals
Certificates
Polysorbate 20, clear cell culture reagent
Supplier:  MP Biomedicals

Specification Test Results

  • Identity Test A:
    Passes
  • Identity Test B:
    Passes
  • FTIR:
    Conforms to Reference
  • Solubility (1g/20ml):
    Clear, Foamy Solution
  • Specific Gravity:
    1.08-1.13
  • Water Content:
    ≤3%
  • Saponification Number:
    40-50

Specification Test Results

Frequently Bought Together