Polysorbate 20, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:9005-64-5
:
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986
Density:1.1 g/cm³ (20 °C)
Flash Pt:149 °C (closed cup)
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to brown-yellow
- IR Spectrum:Passes test
- Density (20/4):1.095 - 1.105
- Hydroxyl value:96 - 113
- Saponification value:40 - 51
Specification Test Results
Frequently Bought Together