Calcium-D-gluconate monohydrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:66905-23-5
:299-28-5
Formula:C₁₂H₂₂CaO₁₄·H₂O
MDL Number:MFCD00150787
:448.39 g/mol
Merck Index:14,1670
Melting Pt:∼ 195 °C (dec.)
Storage Temperature:Ambient
EINECS:206-075-8
Related Information
Frequently Bought Together