Olovo 1.000 mg/l v zřed. kyselině dusičné (from Pb(NO3)2) ARISTAR® standard pro ICP
Supplier: VWR Chemicals
Certificates
About this item
Pb(NO₃)₂ in 2% HNO₃.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.
Limited Time Sale
Select items below are currently on sale
Related Information
Certificates
Search by Lot Number125 ml Plastic bottle for liquids DIN 24 (ZPACK15150125) 500 ml Plastic bottle for liquids DIN 28 (ZPACK15150500)
Packaging Information
VWR® for Calibrations
Product Literature
Frequently Bought Together