Xanthine oxidase (from buttermilk)
Supplier: Merck
Certificates
About this item
Xanthine oxidase from Buttermilk, grade III.
Specifications
- CAS No.:9002-17-9
- Source:Buttermilk
- Enzyme name:Xanthine oxidase
Specifications
Frequently Bought Together