Platinum rhodium alloy (90:10 w%), wire for ISA type S standard grade thermocouple, Ø 0.076 mm (0.003 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
MDL Number:MFCD00198186
Storage Temperature:Ambient
Frequently Bought Together