ContactUsLinkComponent
Platinum rhodium alloy (87:13 w%), wire for ISA type R standard grade thermocouple, Ø 0.127 mm (0.005 in)
Platinum rhodium alloy (87:13 w%), wire for ISA type R standard grade thermocouple, Ø 0.127 mm (0.005 in)
Certificates
Platinum rhodium alloy (87:13 w%), wire for ISA type R standard grade thermocouple, Ø 0.127 mm (0.005 in)