Platinum iridium alloy (80:20 w%) ≥99.9% (metals basis excluding precious metals), wire, hard, Ø 0.152 mm (0.006 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
MDL Number:MFCD02091683
Storage Temperature:Ambient
Frequently Bought Together