Order Entry
Czech Republic
ContactUsLinkComponent
Nickel chromium alloy (80:20 w%), wire, Ø 1.6 mm (0.064 in)
Health
Irritant
Nickel chromium alloy (80:20 w%), wire, Ø 1.6 mm (0.064 in)
Catalog # 42838.G6
CAS Number:  
Health
Irritant
Nickel chromium alloy (80:20 w%), wire, Ø 1.6 mm (0.064 in)
Catalog # 42838.G6
CAS Number:  

Specifications

  • Pk:
    10 m
  • Seq:
    8390

Specifications

About this item

 :
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:11106-97-1
MDL Number:MFCD00673110