ContactUsLinkComponent
Nickel chromium alloy (80:20 w%) ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Nickel chromium alloy (80:20 w%) ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
  40870.KS
 :  
Nickel chromium alloy (80:20 w%) ≥99.95% (metals basis), sputtering target, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
  40870.KS
 :  
  • Pk:
    1
  • Seq:
    0001
 :
 :GHS08
 :GHS07
 :Ambient
 :11106-97-1
 :MFCD00673110