
Lactic acid ≥85% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:50-21-5
:598-82-3
Formula:H₃CCH(OH)COOH
MDL Number:MFCD00004520
Boiling Pt:122 °C (20 hPa)
Density:1.11…1.21 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
:90.08 g/mol
EINECS:200-018-0
:GHS05
Storage Temperature:Ambient
Frequently Bought Together