Order Entry
Czech Republic
ContactUsLinkComponent
 
Starch, soluble for analysis
 
undefined
Starch, soluble for analysis
 

 

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤10 %
  • Sulfated ash:
    ≤0.5 %
  • Identification:
    Positive
  • pH:
    5 to 7.5 (2 % at 25°C)
  • Sensitivity:
    Passes test

 

 

Frequently Bought Together