Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
Agar

Agar

Plastic bottle for solids, 250 g
Catalog # USBIA0950
Supplier:   US Biological
Agar
Catalog # USBIA0950
Supplier:  US Biological
CAS Number:  9002-18-0
Agar
Agar

Plastic bottle for solids, 250 g

CAS Number:  9002-18-0
Price
10 380,00 Kč
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.