Order Entry
Czech Republic
ContactUsLinkComponent
 
Agar, fine powder, EMPROVE® ESSENTIAL Ph. Eur., BP, extra pure, SAFC®
Catalog # 1.01615.1000
CAS Number:  
undefined
Agar, fine powder, EMPROVE® ESSENTIAL Ph. Eur., BP, extra pure, SAFC®
Catalog # 1.01615.1000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
Merck Index:13,00184
Melting Pt:90 °C
Storage Temperature:Ambient
EINECS:232-658-1
CAS Number:9002-18-0
MDL Number:MFCD00081288