Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
D-(-)-Fructose, Sigma-Aldrich®

D-(-)-Fructose, Sigma-Aldrich®

Supplier:   Merck
D-(-)-Fructose, Sigma-Aldrich®
Supplier:  Merck
Certificates
D-(-)-Fructose, Sigma-Aldrich®
D-(-)-Fructose, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
Synonyms:  D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
  • CAS Number:57-48-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148910
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04295
  • Boiling Pt:552 °C (1013 hPa)
  • Melting Pt:100…110 °C
  • Density:1.59 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:200-333-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 1 650,00 Kč
    1 650,00 Kč
    1 * 100 g
       1 650,00 Kč

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 3 720,00 Kč
    3 720,00 Kč
    1 * 500 g
       3 720,00 Kč

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 6 320,00 Kč
    6 320,00 Kč
    1 * 1 kg
       6 320,00 Kč

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.