Order Entry
Czech Republic
ContactUsLinkComponent
D-(+)-Glucose Reag. Ph. Eur., Millipore®
D-(+)-Glucose Reag. Ph. Eur., Millipore®
Catalog # 1.08337.0250
Supplier:  Merck
CAS Number:  
undefined
D-(+)-Glucose Reag. Ph. Eur., Millipore®
Catalog # 1.08337.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
Merck Index:14,04459
Melting Pt:146 °C
Density:1.54 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
 :180.16 g/mol
Storage Temperature:Ambient
EINECS:200-075-1
CAS Number:50-99-7
 :492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912