Order Entry
Czech Republic
ContactUsLinkComponent
Deuterium chloride 20% (w/w) in D₂O 99+ atom % D for NMR spectroscopy
Corrosive
Irritant
Deuterium chloride 20% (w/w) in D₂O 99+ atom % D for NMR spectroscopy
Catalog # ACRO175420500
CAS Number:  
undefined
Corrosive
Irritant
Deuterium chloride 20% (w/w) in D₂O 99+ atom % D for NMR spectroscopy
Catalog # ACRO175420500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Solution
  • D-Enrichment:
    ≥99.0 %
  • Concentration:
    20 to 22 % w/w
  • Ash:
    ≤0.3 % (1000°C, 0.5 h)

Specification Test Results

About this item

 :
 :37.45
Density:1.2 g/cm³
Storage Temperature:Ambient
EINECS:231-715-8
UN:1789
ADR:8,III
 :GHS05
 :GHS07
CAS Number:7698-05-7
Formula:DCl
MDL Number:MFCD00044642