Deuterium bromide 47% (w/w) in D₂O (99% D) (Isotopic), Sigma-Aldrich®
Catalog # SIAL176710-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:81.9
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:236-894-6
CAS Number:13536-59-9
Formula:DBr
MDL Number:MFCD00044232