D-(+)-Sucrose, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:57-50-1
:
Formula:C₁₂H₂₂O₁₁
MDL Number:MFCD00006626
Merck Index:13,08966
Melting Pt:169…170 °C
Density:1.29 g/cm³ (20 °C)
:342.3 g/mol
Storage Temperature:Ambient
EINECS:200-334-9
Specification Test Results
- Acidity:Max. 0.005 meq/g
- Specific optical rotation (26 %; water):66.4 - 66.8 °
- Glucose:Max. 0.05 %
- Heavy metals (as Pb):Max. 10 ppm
- Ignition residue (SO4):Max. 80 ppm
- Insolubility in water:Max. 30 ppm
- Loss on drying (100°C):Max. 0.05 %
- Reducing sugars (calculated as C6H12O6):Max. 0.1 %
- Cl (Chloride):Max. 5 ppm
- N (Nitrogen):Max. 20 ppm
- SO3 (as SO2):Max. 15 ppm
- SO4 (Sulphate):Max. 20 ppm
- Ba (Barium):Max. 0.02 %
- Ca (Calcium):Max. 10 ppm
- Cd (Cadmium):Max. 0.05 ppm
- Cu (Copper):Max. 0.1 ppm
- Fe (Iron):Max. 0.5 ppm
- Ni (Nickel):Max. 0.1 ppm
- Pb (Lead):Max. 0.5 ppm
- Zn (Zinc):Max. 0.5 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 6L Plastic bottle for solids DIN 80 (ZPACK13256000) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
27480_D(+)-SACCHAROSE ANALAR NORMAPUR_BSE_REV0.pdf
BSE/TSE Certificates
Avantor Cell Culture Solutions.pdf
Product Literature
Frequently Bought Together