D-(+)-Sucrose, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:57-50-1
:
Formula:C₁₂H₂₂O₁₁
MDL Number:MFCD00006626
Merck Index:13,08966
Melting Pt:169…170 °C
Density:1.29 g/cm³ (20 °C)
:342.3 g/mol
Storage Temperature:Ambient
EINECS:200-334-9
Specification Test Results
- Specific optical rotation (2 %; water):66 - 67 °
- Heavy metals (as Pb):Max. 50 ppm
- Ignition residue (SO4):Max. 0.2 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 30L Plastic bucket UN Z (ZPACK13247033) 33L Plastic bucket UN Z (ZPACK13247037) 6L Plastic bottle for solids DIN 80 (ZPACK13256000)
Packaging Information
27478_D(+)-SACCHAROSE GPR RECTAPUR_BSE_REV0.pdf
BSE/TSE Certificates
Avantor Cell Culture Solutions.pdf VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together