ContactUsLinkComponent
D-(+)-Sucrose ≥99.9% (by HPLC), Ultrapure Bioreagent for density gradient ultracentrifugation, for molecular biology, J.T.Baker®
D-(+)-Sucrose ≥99.9% (by HPLC), Ultrapure Bioreagent for density gradient ultracentrifugation, for molecular biology, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
D-(+)-Sucrose ≥99.9% (by HPLC), Ultrapure Bioreagent for density gradient ultracentrifugation, for molecular biology, J.T.Baker®
Supplier:  Avantor
Certificates
Specification Test Results
  • DNase Activity (None Detected):
    Passes Test
  • RNase Activity:
    Passes Test
  • Protease Activity:
    Passes Test
  • Clarity and Color of Solution:
    Passes Test
  • Clarity of Solution:
    Passes Test
  • Glucose:
    ≤0.1 %
  • Total of Copper, Iron and Lead:
    ≤5 ppm
  • Assay (C₁₂H₂₂O₁₁) (by HPLC):
    ≥99.9 %