Zadání objednávky
Czech Republic
ContactUsLinkComponent
 
D(+)-Sacharóza 99+% for biochemistry DNase-, RNase-, protease-free
 
undefined
D(+)-Sacharóza 99+% for biochemistry DNase-, RNase-, protease-free

 

  • Appearance:
    White Crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.03 %
  • Solubility:
    (10 % in water) Clear colorless
  • HPLC:
    ≥99.0 % (ELSD)
  • Dnase,Rnase,Protease:
    None detected
  • Heavy metals (as Pb):
    ≤5 ppm

 

 

Frequently Bought Together