D-(+)-Raffinose pentahydrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:17629-30-0
:
Formula:C₁₈H₃₂O₁₆·5H₂O
MDL Number:MFCD00071590
:594.52 g/mol
Merck Index:14,8096
Melting Pt:78…82 °C
Density:1.516 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-146-9
Related Information
Frequently Bought Together