D-(+)-Glucose monohydrate
Catalog # ICNA0219901390
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:198.17 g/mol
Melting Pt:83 °C
Storage Temperature:Ambient
EINECS:200-075-1
CAS Number:14431-43-7
:5996-10-1
Formula:C₆H₁₂O₆·H₂O
MDL Number:MFCD00149450