D-Gluconic acid 49 - 53% (w/w) in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:526-95-4
:MFCD00066366
Formula:C₆H₁₂O₇
MDL Number:MFCD00004240
:196.16 g/mol
Density:1.24 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-401-4
UN:3265
ADR:8,III
Frequently Bought Together