Order Entry
Czech Republic
ContactUsLinkComponent
 
D-Gluconic acid 50% in aqueous solution for synthesis, Sigma-Aldrich®
 :  Merck
 
D-Gluconic acid 50% in aqueous solution for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    (20 °C) soluble
  • Boiling Point:
    105 - 106 °C (1013 hPa)
  • Density:
    1.24 g/cm3 (20 °C)
  • pH:
    2.2 (500 g/l, H2O, 20 °C)

 

Frequently Bought Together