Order Entry
Czech Republic
ContactUsLinkComponent
D-(+)-Glucose monohydrate
D-(+)-Glucose monohydrate
Catalog # A1349.5000
Supplier:  PanReac AppliChem
CAS Number:  
undefined
D-(+)-Glucose monohydrate
Catalog # A1349.5000
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    5 kg
  • Seq:
    4114

Specifications

About this item

 :
 :198.17 g/mol
Melting Pt:83 °C
Storage Temperature:Ambient
EINECS:200-075-1
CAS Number:14431-43-7
 :5996-10-1
Formula:C₆H₁₂O₆·H₂O
MDL Number:MFCD00149450