D(+)-Maltóza monohydrát, Sigma-Aldrich®
Catalog # SIALM2250-1KG
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:1 kg
- Sekvence:4024
Specifications
About this item
:
Merck Index:13,05736
Melting Pt:160…165 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-716-5
CAS Number:6363-53-7
Formula:C₁₂H₂₂O₁₁·1H₂O
MDL Number:MFCD00149343