ContactUsLinkComponent
D-(+)-Maltose monohydrate 92+%
D-(+)-Maltose monohydrate 92+%
Certificates
undefined
D-(+)-Maltose monohydrate 92+%
Certificates
Specification Test Results
  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤7 %
  • Sulfated ash:
    ≤0.1 %
  • Additional info:
    remainder mainly glucose and maltotriose
  • HPLC:
    ≥92.0 %
  • Specific optical rotation:
    +135° to +139° (20°C, 589 nm) (c=4, H2O, NH3) (on dry substance)