Order Entry
Czech Republic
ContactUsLinkComponent
Boron trifluoride dibutyl etherate, Sigma-Aldrich®
Corrosive
Toxic
Boron trifluoride dibutyl etherate, Sigma-Aldrich®
Catalog # SIAL307912-100ML
Supplier:  Merck
CAS Number:  
Corrosive
Toxic
Boron trifluoride dibutyl etherate, Sigma-Aldrich®
Catalog # SIAL307912-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
Boiling Pt:126 °C (1013 hPa)
Melting Pt:–14 °C
Density:0.959 g/mL (25 °C) (lit.)
EINECS:209-783-5
ADR:8,II
 :M-Clarity™ quality level = MQ100
Flash Pt:62 - 63 °C
 :198.04 g/mol
Storage Temperature:Ambient
UN:3264
 :GHS05
 :GHS06
CAS Number:593-04-4
Formula:BF₃·C₈H₁₈O
MDL Number:MFCD00013195