



Fluorid boritý - ether komplex, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:109-63-7
:
Formula:C₄H₁₀BF₃O
MDL Number:MFCD00013194
Merck Index:13,01340
Boiling Pt:126 °C (1013 hPa)
Melting Pt:–60 °C
Density:1,125 g/cm³ (25 °C)
Flash Pt:59 °C
UN:2604
ADR:8,I
:M-Clarity™ quality level = MQ200
EINECS:203-689-8
:GHS02
:GHS06
:GHS05
:GHS08
Storage Temperature:Ambient
Frequently Bought Together