Dimethyl sulphoxide-[D₆] (99.5% D) for NMR spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2206-27-1
:
Formula:D₃CS(O)CD₃
MDL Number:MFCD00002090
:84.09 g/mol
Boiling Pt:190 °C (760 mmHg)
Melting Pt:20.2 °C
Density:1.19 g/cm³
Flash Pt:88 °C
Storage Temperature:Ambient
EINECS:218-617-0
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.05 % (Coulometric)
- D-Enrichment:≥99.50 %
Specification Test Results
Frequently Bought Together