Order Entry
Czech Republic
ContactUsLinkComponent
Toluene-[D₈], Sigma-Aldrich®
Flammable
Health
Irritant
Toluene-[D₈], Sigma-Aldrich®
Catalog # SIAL570710-50G
Supplier:  Merck
CAS Number:  
Flammable
Health
Irritant
Toluene-[D₈], Sigma-Aldrich®
Catalog # SIAL570710-50G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :100.08 g/mol
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–85 °C
Density:0.94 g/cm³ (20 °C)
Flash Pt:4 °C (closed cup)
Storage Temperature:Refrigerator
EINECS:218-009-5
UN:1294
ADR:3,II
 :M-Clarity™ quality level = MQ100
 :GHS02
 :GHS08
 :GHS07
CAS Number:2037-26-5
Formula:C₆D₅CD₃
MDL Number:MFCD00044638