


Methanol-[D₁], Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1455-13-6
:
Formula:CH₃DO
MDL Number:MFCD00044213
:33.03 g/mol
Boiling Pt:64.5 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.81 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
UN:1230
ADR:3,II
:M-Clarity™ quality level = MQ100
EINECS:215-933-0
:GHS02
:GHS06
:GHS08
Frequently Bought Together