

2-Propanol-[D₈], Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:22739-76-0
:
Formula:D₃CCD(OD)CD₃
MDL Number:MFCD00044341
:68.03 g/mol
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89.5 °C
Density:0.89 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:245-189-2
ADR:3,II
:M-Clarity™ quality level = MQ200
UN:1219
:GHS02
:GHS07
Frequently Bought Together