Order Entry
Czech Republic
ContactUsLinkComponent
2-Propanol-[D7]
Flammable
Irritant
2-Propanol-[D7]
Catalog # 8.15046.0001
Supplier:  Merck
CAS Number:  
Flammable
Irritant
2-Propanol-[D7]
Catalog # 8.15046.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :67.04 g/mol
Storage Temperature:Ambient
UN:1219
ADR:3,II
 :GHS02
 :GHS07
CAS Number:19214-96-1
Formula:D₃CCD(OD)CD₃