

Diethyl ether-[D₁₀], Sigma-Aldrich®
Catalog # SIAL613479-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:84.04 g/mol
Boiling Pt:34.6 °C (1013 hPa)
Melting Pt:–116.3 °C
Density:0.714 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:220-235-4
UN:1155
ADR:3,I
:M-Clarity™ quality level = MQ100
:GHS02
:GHS07
CAS Number:2679-89-2
Formula:D₃CCD₂OCD₂CD₃
MDL Number:MFCD00062316